missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ 80 Position Fraction Collection Plate

Product Code. p-7221529 Shop All Gilson Products
Click to view available options
Quantity:
100 Pk.
20 Pk.
Unit Size:
Pack of 100
Pack of 20
This item is not returnable. View return policy

Product Code. 16606115

Brand: Gilson™ 12240102

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These accessories are for use with Gilson fraction collectors.

  • Includes drop detector heads, dispense needles, collection plates, and a multiple column adapter
  • Vessel Type is Cavities
  • Purification Technique is HPLC

Specifications

For Use With (Equipment) 2.5 mL Cavities
Quantity 20 Pk.
No. of Positions 80
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.