Learn More
Moldex™ 8000 Series Half-Mask Respirators Cartridges
For use with 8000 series half mask respirators
Brand: Moldex™ 810001
Code : S8
Additional Details : Weight : 0.01000kg
Description
- Use with 8910 N95, 8970 R95 or 8940 P100 filters against particulates
- The P100 cartridge can be used for both oil and non-oil based particulates
- 100% PVC-Free™
Specifications
Organic Vapor Cartridge | |
Organic Vapor Cartridge | |
Series 8000 half mask, n95, r95, p100 filter |
NIOSH | |
For protection against organic vapors | |
10/Pk. |