missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid
Hold eight-one 1.5 or 2.0mL cryogenic storage vials with this stackable cryogenic storage rack with lid.
Brand: Thermo Scientific™ 8830-11
10.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Hold eighty-one 1.5 or 2.0mL cryogenic storage vials with the Thermo Scientific™ 81-Well Cryogenic Storage Rack with Lid. Available in several colors with transparent tinted lid that protects samples from dust and debris and permits stacking.
- Molded with alphanumeric grid for easy tube identification
- Lid protects samples and allows racks to be stacked
Specifications
Fluorescent Pink | |
81 (1.5 mL or 2.0 mL Tubes) | |
Polypropylene | |
81-Well Cryogenic Storage Racks with Lid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction