Learn More
Cytiva 850-DS PVDF 8-Channel Filter Plates
Used for filtering purposes.
Brand:  Cytiva 7707-3800
Additional Details : Weight : 1.70000kg
 View more versions of this product
Product Code. 15661067
  • £545.00 / Pack of 50
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
Description
Description
  • Used for filtering purposes/li>
Specifications
Specifications
PVDF
Filtering
850-DS PVDF 8-Channel Filter Plates
0.45 μm
50/Pk.