missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 8mm Aluminum Crimp Seal, Silver, Center hole, Assembled septum

Provide a secure leak-resistant seal.

Brand:  Fisherbrand™ 08 03 2042

 View more versions of this product

Product Code. 11818981

  • £84.30 / Pack of 1000
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Aluminum crimp seals with prefitted septa
  • Provide a secure leak-resistant seal
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Silicone/PTFE
Beige, Red
Aluminum
100
Crimp Cap
4 mm
8 mm
GC, HPLC
1 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers