missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 8mm Glass Crimp Neck Vial

Can be closed with 8mm Aluminum Caps, 9mm PE-Caps or with 8mm Push-On Caps.

Brand:  Fisherbrand™ 08 09 0305

 View more versions of this product

Product Code. 11782408

  • £471.00 / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Available in different volumes
  • Flat, round or conical bottom
  • Clear or amber glass
  • For almost all autosamplers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Micro-Vial
7 mm
0.64 mL
Crimp
<3 μL
Conical Bottom
100
No
Clear
Glass
GC, HPLC
CleanPack
0.6 mL
40 mm
ND8
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ 8mm Glass Crimp Neck Vial > Clear, conical, 0.6mL, 40mm height

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.