missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 8mm PP Screw Cap, 8-425

Polypropylene caps are chemically inert and suitable for most chromatography applications.

Brand:  Fisherbrand™ 08 08 0436

 View more versions of this product

Product Code. 11898981

  • £62.05 / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • 8mm
  • PP
  • Screw Cap
  • 8-425 thread
100 pcs. per pack
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
Polypropylene
Center Hole Screw Cap
8 mm
100
ND8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.