missing translation for 'onlineSavingsMsg'
Learn More
9,10-Dichloroanthracene, 97%, Thermo Scientific Chemicals Product Code.: 11475203

9,10-Dichloroanthracene, 97%, Thermo Scientific Chemicals

Product Code. 11475203
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11475203

Brand: Thermo Scientific Alfa Aesar A12823.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-48-1
Molecular Formula C14H8Cl2
Molecular Weight (g/mol) 247.118
MDL Number MFCD00001246
InChI Key FKDIWXZNKAZCBY-UHFFFAOYSA-N
PubChem CID 11800
IUPAC Name 9,10-dichloroanthracene
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=C2Cl)Cl

Specifications

Melting Point 210°C to 215°C
Quantity 5 g
Beilstein 1912108
Synonym anthracene, 9,10-dichloro, acmc-209mkk, anthracene,10-dichloro, 9,10-dichloro-anthracene, anthracene,9,10-dichloro, 9.10-dichloroanthracene
Formula Weight 247.12
Percent Purity 97%
Chemical Name or Material 9,10-Dichloroanthracene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252748 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberA12823.14,A12823.06
Lot Number10198705 Certificate TypeCertificate of Analysis Date10/09/2024 Catalog NumberA12823.14,A12823.06
Lot Number10238399 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA12823.14,A12823.06
Lot Number10188252 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12823.14,A12823.06
Lot Number10160393 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12823.14,A12823.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9,10-Dichloroanthracene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.