missing translation for 'onlineSavingsMsg'
Learn More
9,10-Diphenylanthracene, 99%, Thermo Scientific Chemicals Product Code.: 11487687

9,10-Diphenylanthracene, 99%, Thermo Scientific Chemicals

Product Code. 11487687
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11487687

Brand: Thermo Scientific Alfa Aesar A15714.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.

Solubility
Soluble in ethanol, chloroform, benzene and carbon disulfide. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-10-1
Molecular Formula C26H18
Molecular Weight (g/mol) 330.43
MDL Number MFCD00001253
InChI Key FCNCGHJSNVOIKE-UHFFFAOYSA-N
Synonym anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standardShow MoreShow Less
PubChem CID 15159
ChEBI CHEBI:51676
IUPAC Name 9,10-diphenylanthracene
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 248°C to 254°C
Odor Odorless
Quantity 1 g
Beilstein 1914010
Solubility Information Soluble in ethanol,chloroform,benzene and carbon disulfide. Insoluble in water.
Formula Weight 330.43
Percent Purity 99%
Chemical Name or Material 9,10-Diphenylanthracene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS16I039 Certificate TypeCertificate of Analysis Date22/06/2022 Catalog NumberA15714.03,A15714.14,A15714.06
Lot NumberR31F018 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15714.03,A15714.14,A15714.06
Lot NumberP24A034 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15714.03,A15714.14,A15714.06
Lot NumberE23T025 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15714.03,A15714.14,A15714.06
Lot NumberL20W037 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15714.03,A15714.14,A15714.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9,10-Diphenylanthracene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.