missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-octylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15412076
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 258865-48-4
Molecular Formula C29H44B2O4
Molecular Weight (g/mol) 478.29
MDL Number MFCD03701608
InChI Key HURJMQMZDPOUOU-UHFFFAOYSA-N
Synonym 9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid
PubChem CID 4283240
IUPAC Name (7-borono-9,9-dioctylfluoren-2-yl)boronic acid
SMILES CCCCCCCCC1(CCCCCCCC)C2=CC(=CC=C2C2=C1C=C(C=C2)B(O)O)B(O)O

Specifications

Quantity 250 mg
Formula Weight 478.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 9,9-Di-n-octylfluorene-2,7-diboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'