missing translation for 'onlineSavingsMsg'
Learn More
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%, Thermo Scientific Chemicals Product Code.: 15479465

9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%, Thermo Scientific Chemicals

Product Code. 15479465
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15479465

Brand: Thermo Scientific Alfa Aesar H51955.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester is a reactant in the synthesis of 6,6-(5,5-(9,9-dioctyl-9H-fluorene-2,7-diyl)bis(thiophene-5,2-diyl))bis(2,5-bis(2-ethylhexyl)-3-(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione) (DPP1), a solution-processable nonfullerene electron acceptor used in organic solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester is a reactant in the synthesis of 6,6-(5,5-(9,9-dioctyl-9H-fluorene-2,7-diyl)bis(thiophene-5,2-diyl))bis(2,5-bis(2-ethylhexyl)-3-(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione) (DPP1), a solution-processable nonfullerene electron acceptor used in organic solar cells.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196207-58-6
Molecular Formula C41H64B2O4
Molecular Weight (g/mol) 642.579
MDL Number MFCD16294554
InChI Key FAHIZHKRQQNPLC-UHFFFAOYSA-N
Synonym 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-di-n-octylfluorene, 9,9-di-n-octylfluorene-2,7-diboronic acid bis pinacol ester, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dioctylfluorene, 2-9,9-dioctyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-9,9-dioctyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb260, 9,9-dioctylfluorene-2,7-bis boronic acid pinacol ester, 9,9-dioctylfluorene-2,7-diboronic acid bis pinacol ester, 9,9-dioctyl-9h-fluorene-2,7-diboronic acid bis pinacol esterShow MoreShow Less
PubChem CID 21982074
IUPAC Name 2-[9,9-dioctyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(C3(CCCCCCCC)CCCCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 642.58
Percent Purity 95%
Chemical Name or Material 9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW07J018 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog NumberH51955.06,H51955.03
Lot NumberB10X047 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51955.06,H51955.03
Lot NumberW22H019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51955.06,H51955.03
Lot NumberN02H014 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51955.06,H51955.03
Lot NumberR13C023 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51955.06,H51955.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.