missing translation for 'onlineSavingsMsg'
Learn More

9-Aminofluorene hydrochloride, 98+%

Product Code. 11311847
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11311847

Brand: Thermo Scientific Alfa Aesar L05156.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5978-75-6 | C13H12ClN | 217.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5978-75-6
Molecular Formula C13H12ClN
Molecular Weight (g/mol) 217.70
MDL Number MFCD00012536
InChI Key SYKJOJSYQSVNOM-UHFFFAOYSA-N
Synonym 9h-fluoren-9-amine hydrochloride, 9-aminofluorene hydrochloride, 9h-fluoren-9-amine, hydrochloride, 9-fluorenamine hydrochloride, 9-aminofluorene hcl, fluorene-9-ylamine, chloride, 9h-fluoren-9-amine hcl, acmc-1b10k, 9-aminoflu-orene hydrochloride, 9-aminofluorene monohydrochloride
PubChem CID 2724685
SMILES [H+].[Cl-].NC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point ∼225°C
Quantity 1 g
Beilstein 3915368
Sensitivity Hygroscopic
IUPAC Name 9H-fluoren-9-amine;hydrochloride
Formula Weight 217.7
Percent Purity ≥98%
Chemical Name or Material 9-Aminofluorene hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.