missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Benzylcarbazole-3-boronic acid pinacol ester, 95%, Thermo Scientific™
£31.00 - £99.30
Chemical Identifiers
CAS | 1357387-29-1 |
---|---|
Molecular Formula | C25H26BNO2 |
Molecular Weight (g/mol) | 383.298 |
MDL Number | MFCD16294547 |
InChI Key | UMWJZMUSYJMTOI-UHFFFAOYSA-N |
Synonym | 9-benzyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-9h-carbazole, 9-benzyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9-benzyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl carbazole, amtb246, 9benzylcarbazole-3-boronic acid pinacol ester, 9-benzylcarbazole-3-boronic acid pinacol ester, 9-benzyl-9h-carbazole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9-benzyl-9h-carbazole |
PubChem CID | 71306536 |
IUPAC Name | 9-benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=CC=CC=C43)CC5=CC=CC=C5 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1357387-29-1 | |
383.298 | |
UMWJZMUSYJMTOI-UHFFFAOYSA-N | |
71306536 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=CC=CC=C43)CC5=CC=CC=C5 |
C25H26BNO2 | |
MFCD16294547 | |
9-benzyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-9h-carbazole, 9-benzyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9-benzyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl carbazole, amtb246, 9benzylcarbazole-3-boronic acid pinacol ester, 9-benzylcarbazole-3-boronic acid pinacol ester, 9-benzyl-9h-carbazole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9-benzyl-9h-carbazole | |
9-benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole |
Specifications
1357387-29-1 | |
MFCD16294547 | |
UMWJZMUSYJMTOI-UHFFFAOYSA-N | |
9-benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole | |
71306536 | |
95% |
C25H26BNO2 | |
9-benzyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-9h-carbazole, 9-benzyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9-benzyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl carbazole, amtb246, 9benzylcarbazole-3-boronic acid pinacol ester, 9-benzylcarbazole-3-boronic acid pinacol ester, 9-benzyl-9h-carbazole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9-benzyl-9h-carbazole | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=CC=CC=C43)CC5=CC=CC=C5 | |
383.298 | |
383.3 | |
9-Benzylcarbazole-3-boronic acid pinacol ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures