missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-beta-D-Arabinofuranosyladenine-5'-monophosphate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Disrupts viral DNA synthesis. Competitive viral DNA polymerase inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Disrupts viral DNA synthesis. Competitive viral DNA polymerase inhibitor. Active against herpes simplex and varicella zoster viruses in vivo.
Solubility
Soluble in water at 4mg/ml
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Disrupts viral DNA synthesis. Competitive viral DNA polymerase inhibitor. Active against herpes simplex and varicella zoster viruses in vivo.
Solubility
Soluble in water at 4mg/ml
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Specifications
Specifications
Color | White |
Quantity | 5 g |
Solubility Information | Soluble in water at 4mg/ml |
Formula Weight | 347.22 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | 9-beta-D-Arabinofuranosyladenine-5'-monophosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'