missing translation for 'onlineSavingsMsg'
Learn More

9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 165811000

53.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.



 View more versions of this product

Product Code. 10315092

  • £71.49 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid, Reduction reagent
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465251 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number165811000,165818000
Lot NumberA0461493 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number165811000,165818000
Lot NumberA0453590 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number165811000,165818000
Lot NumberA0443943 Certificate TypeCertificate of Analysis Date19/01/2023 Catalog Number165811000,165818000
Lot NumberA0403367 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number165811000,165818000
Special Offers

Special Offers

  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    21

    Minutes

    38

    Seconds

    37
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals > 100mL; AcroSeal glass bottle packed in tin can

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only