missing translation for 'onlineSavingsMsg'
Learn More

9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 165818000

263.64 GBP valid until 2024-12-31
Use promo code "21665" to get your promotional price.



 View more versions of this product

Product Code. 10609834

  • £335.67 / 800mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid, Reduction reagent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

9-Borabicyclo[3.3.1]nonane
6.14, 92.49
0.8840g/mL
AcroSeal™ Glass Bottle
MFCD00074742
02,31; 03,24; 04,41; 05,46; 06,62; 07,29; 08,47; 09,57; 10,48; 11,68; 13,92; 14,52; 15,43; 17,49; 01,732; 04,356
9-borabicyclo 3.3.1 nonane, 9-borabicyclo 3.3.1 nonan, 9-borabicyclo 3,3,1 nonane, 9$l^ 2-borabicyclo 3.3.1 nonane, 1s,5s-9-borabicyclo 3.3.1 nonane, monomer; 21205-91-4 dimer, 9-borabicyclo 3.3.1 nonane,9-bbn,intetrahydrofuran
AMKGKYQBASDDJB-UHFFFAOYSA-N
9$l^{2}-borabicyclo[3.3.1]nonane
6327450
280-64-8, 109-99-9
7.51, 93.86
−17°C
C8H15B
800 mL
0.884
Solubility in water: reacts
[B]1C2CCCC1CCC2
122.02
122.02
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H261-In contact with water releases flammable gases.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 9-Borabicyclo[3.3.1]nonane, Tetrahydrofuran

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only