missing translation for 'onlineSavingsMsg'
Learn More

9-Chloroanthracene, 96%, Thermo Scientific Chemicals

Product Code. 11334454
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 716-53-0
Molecular Formula C14H9Cl
Molecular Weight (g/mol) 212.676
MDL Number MFCD00001245
InChI Key KULLJOPUZUWTMF-UHFFFAOYSA-N
PubChem CID 69744
IUPAC Name 9-chloroanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Cl

Specifications

Melting Point 101°C to 106°C
Quantity 25 g
Beilstein 1869540
Synonym anthracene, 9-chloro, ccris 5547, 9-chloro-anthracene, #, anthracene,9-chloro, bidd:gt0730
Solubility Information Insoluble in water.
Formula Weight 212.68
Percent Purity 96%
Chemical Name or Material 9-Chloroanthracene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'