missing translation for 'onlineSavingsMsg'
Learn More

9-Dodecyn-1-ol, 97%, Thermo Scientific™

Product Code. 11485147
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71084-08-7
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.307
MDL Number MFCD00082940
InChI Key MIDXCKJJIFQOLT-UHFFFAOYSA-N
Synonym 9-dodecyn-1-ol
PubChem CID 117011
IUPAC Name dodec-9-yn-1-ol
SMILES CCC#CCCCCCCCCO

Specifications

Linear Formula CH3CH2C≡C(CH2)8OH
Refractive Index 1.4625
Quantity 5 g
Beilstein 6642771
Formula Weight 182.31
Percent Purity 97%
Chemical Name or Material 9-Dodecyn-1-ol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'