missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenylmethyl carbamate, 99%, Thermo Scientific Chemicals

Product Code. 10442770
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84418-43-9
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.28
MDL Number MFCD00237376
InChI Key ZZOKVYOCRSMTSS-UHFFFAOYSA-N
Synonym 9-fluorenylmethyl carbamate, fmoc-nh2, fmoc-amide, 9-fluorenylmethylcarbamate, n-9-fluorenylmethoxycarbonyl amide, fmoc amine, fmoc-amine, 9-fluorenylcarbamate, 9-fluorenemethane resin, pubchem12066
PubChem CID 736301
IUPAC Name 9H-fluoren-9-ylmethyl carbamate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 204°C to 207°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 239.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 9-Fluorenylmethyl carbamate
Show More Show Less
compliance-icons
Product Identifier
  • 9-Fluorenylmethyl carbamate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'