missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 9 mm Short Thread TPX Vial with Integrated Glass Micro-Insert, Silanized
64.75 GBP valid until 2025-03-14
Use promo code "24142" to get your promotional price.
Description
- Universally compatible on almost all autosamplers, thereby rationalization of other 1.5 mL vials, as for instance 11 mm Crimp Neck Vials, Screw Neck Vials 8-425 and 10-425, is possible
- Vials with integrated Micro-Insert are available in amber glass

Specifications
TopSert | |
11.6 mm | |
0.36 mL | |
Short Thread | |
Conical Integrated Insert | |
0.2 mL | |
32 mm | |
Autosampler Vial | |
No |
Amber | |
TPX | |
GC, HPLC | |
CleanPack, Polypropylene Box | |
<1 μL | |
Flat Bottom | |
100 | |
ND9 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Incredible Savings on Peptide and Protein Mass Spectrometry Testing Products
Explore our new Peptide and Protein Mass Spectrometry Testing Products PromotionPromo Code: 24142Expires: 2025-03-14
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
16Hours
22Minutes
14Seconds
24
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ 9 mm Short Thread TPX Vial with Integrated Glass Micro-Insert, Silanized > Amber, silanized
Spot an opportunity for improvement?Share a Content Correction