missing translation for 'onlineSavingsMsg'
Learn More

9-Phenyl-2,3,7-trihydroxy-6-fluorone, Thermo Scientific Chemicals

Product Code. 11460403
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Phenyl-2,3,7-trihydroxy-6-fluorone is used in presence of cetylpyridinium chloride for spectrophotometric determination of copper(II) in blood serum. It is used in the determination of germanium. It is also used to determine molybdenum(VI).

Solubility
Soluble in chloroform.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 975-17-7
Molecular Formula C19H12O5
Molecular Weight (g/mol) 320.3
MDL Number MFCD00005048
InChI Key YDCFOUBAMGLLKA-UHFFFAOYSA-N
Synonym phenylfluorone, fluorone black, 9-phenyl-2,3,7-trihydroxy-6-fluorone, fluorone, phenyl, 2,6,7-trihydroxy-9-phenyl-3h-xanthen-3-one, 9-pheny-3-fluorone, 3h-xanthen-3-one, 2,6,7-trihydroxy-9-phenyl, 9-phenyl-3-fluorone, 2,3,7-trihydroxy-9-phenyl-6-fluorone, 2,6,7-trihydroxy-9-phenyl-3-isoxanthone
PubChem CID 70420
IUPAC Name 2,6,7-trihydroxy-9-phenylxanthen-3-one
SMILES C1=CC=C(C=C1)C2=C3C=C(C(=O)C=C3OC4=CC(=C(C=C42)O)O)O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 40910
Solubility Information Soluble in chloroform.
Formula Weight 320.3
Chemical Name or Material 9-Phenyl-2,3,7-trihydroxy-6-fluorone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'