missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Phenylacridine, Thermo Scientific™
Brand: Thermo Scientific Acros 469430050
This item is not returnable.
View return policy
Description
Specifications
PHENYLACRIDINE | |
602-56-2 | |
Yellow | |
Conforms | |
C19 H13 N | |
5 g | |
Crystalline Powder |
9-Phenylacridine | |
183°C to 187°C | |
411°C | |
Glass bottle | |
00092562 | |
255.32 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction