Learn More
Uvex™ 9188 Cybric Safety Spectacles
Cybric spectacles offer a large field of vision and individual adjustment capabilities for reliable protection and comfort.
Specifications
Lens Type Supravision Sapphire
Frame Color Black, Orange
Model 9188 Cybric
Lens Material Polycarbonate
UV Protection UV400
View More Specs
Description
Description
  • Spectacles feature a large field of vision and individual adjustment capabilities for reliable protection and comfort
  • Certified according to EN 166 (personal eye protection) and EN 170 (UV filters)
  • Quattroflex side arms with four comfort cushions to ensure a comfortable, pressure-free fit around the ear
  • Inclination of the side arms can be adjusted between multiple levels, allowing them to be adapted individually to all facial shapes
  • Supravision sapphire coating is extremely chemical-resistant and scratch-resistant on both sides
  • Reliable UV 400 protection
Specifications
Specifications
Supravision Sapphire
9188 Cybric
UV400
Black, Orange
Polycarbonate