missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 9193 Sportstyle Safety Spectacles

Catalog Number 17277281
10.76 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.


Click to view available options
Lens Tint:
Clear
Gray
Light Brown (CBR65)
Light Green
Silver Mirror
Lens Coating:
Anti-fog

Innovative safety spectacles featuring a sporty design, combined with maximum functionality and safety. Feather-light construction weighs only 23 grams, making these spectacles comfortable to wear for extended periods of time.

  • Innovative safety spectacles feature a sporty design that combines functionality and safety 
  • Feather-light construction weighs only 23 grams
  • Soft, adjustable nosepiece with soft, anti-slip earpieces for a pressure-free fit
  • Lens geometry ensures optimum vision

Supravision AR Coating

  • Inside: Anti-reflective
  • Outside: Anti-reflective
  • Ideal for working conditions with strong or mixed lighting conditions

Supravision Excellence Coating

  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling

Supravision Extreme Coating

  • Interior: Anti-fog
  • Exterior: Scratch-resistant
  • Ideal for working areas with long-term exposure to condensation, and high levels of dirt

Supravision Plus Coating

  • Inside: Anti-fog and scratch-resistant
  • Outside: Anti-fog and scratch-resistant
  • Suitable for environments with frequent temperature and humidity fluctuations

Supravision Variomatic Coating

  • Inside: Anti-fog
  • Outside: Scratch-resistant
  • Self-coloring filters automatically darken within 10 seconds and brighten after 30 seconds
  • Suitable for working areas with frequent movement from inside to outside, and from light to dark
Certifications/Compliance EN 166:2001, EN 172:1994 + A1:2000 + A2:2001
Lens Type Supravision Extreme
Lens Tint Light Brown (CBR65)
Frame Color White, Black
Model 9193 Sportstyle
Weight (Metric) 23 g
Disposable Reusable
Lens Material Polycarbonate
UV Protection UV400
Labeling W 166 FT CE - 5-1.4 W1 FTKN CE
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'