Learn More
uvex 9199 X-Fit Safety Spectacles
Product Code. 17257311 Shop All Uvex Products
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Lens Tint:
Amber
Clear
Gray
Unit Size:
Each
Product Code. 17257311
Brand: uvex 9199280
Only null left
Add to basket
Add to basket
Sporty spectacles with a lightweight design that provides optimum eye coverage, and a high level of user acceptance.
  • Sporty spectacles ensure optimum coverage of the entire eye area
  • Innovative hinged side arm for a personalized fit
  • Lightweight design weighs only 23 grams
  • Metal-free
Supravision Excellence Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
Supravision Sapphire Coating
  • Inside: Scratch-resistant and chemical-resistant
  • Outside: Scratch-resistant and chemical-resistant
  • Non-stick properties allow for easy removal of greasy/watery dirt
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001, EN 172:1994 + A1:2000 + A2:2001
Lens Type Supravision Excellence
Lens Coating Anti-fog, Scratch-resistant
Filter Type Sunglare
Lens Tint Gray
Product Type Ergonomic Safety Glasses
Frame Color Gray
Model 9199 X-Fit
Transmittance Value 0.23
Weight (Metric) 23 g
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Labeling W 166 FT CE - 5-2,5 W 1 FTKN CE