Learn More
Uvex™ 9301 Ultravision Wide-Vision Safety Goggles
12.21 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Lens Type:
Replaceable, with two tear-off films
Description
- Cleverly designed ventilation system helps prevent fogging
- Design is particularly well-suited for use with a respirator
- Wide-vision goggles can be worn comfortably over prescription glasses
- Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
- Supravision excellence coating is extremely scratch-resistant and chemical-resistant on the outside, and anti-fog on the inside
- Lenses can be replaced for extended goggle life
Specifications
Lens Type | Replaceable, with two tear-off films |
Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
Lens Tint | Clear |
Frame Color | Transparent Gray |
Model | 9301 Ultravision (Wide-Vision) |
UV Protection | UV400 |
Labeling | W 166 34 BT CE – 2C-1,2 W 1 BT KN DIN CE |