missing translation for 'onlineSavingsMsg'
Learn More

uvex 95798 Insole For Uvex 1 Sport - Width 11

Product Code. 17258971 Shop All Uvex Products
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
51
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17258971

Brand: uvex 9579836

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Comfortable climatic insole with antistatic properties for Uvex 1 Sport

  • Full-surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No. 9579.8
  • Sizes: 35 to 52
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Uvex 1 Sport
Material Microvelour
Size 36
Disposable Single-use
Gender Unisex
Fit Standard Width
Latex Free Yes
Waterproof No
Product Type Insoles
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.