Learn More
Fisherbrand™ 96 PP Well Plate with 96 Glass Insert and PE Snap Caps with Septum
For complex UHPLC, LC and GC applications
Brand:  Fisherbrand™ 08 20 0911
136.79 GBP valid until 2024-11-30
Use promo code "21756" to get your promotional price.
 View more versions of this product
Product Code. 11531404
  • £147.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 96 Wells
  • Silicone/PTFE
  • Clear
  • Glass
  • Plates meet ANSI and SBS standards
  • Different sealing options
  • Chromatography tested
  • No leeching due to high-end material
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Clear
96
Round
8 x 12
Glass
1200 μL