missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well DeepWell™ Polypropylene Microplates
GREENER_CHOICE

Ideal for storage because of the low-binding characteristics of polypropylene.

Brand:  Fisherbrand™ 260254

113.00 GBP valid until 2024-10-31
Use promo code "21661" to get your promotional price.


 View more versions of this product

Product Code. 13515450

  • £138.00 / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Ideal for storage because of the low-binding characteristics of polypropylene

  • Accommodate even larger working volumes; up to 1.0 mL or 2.0 mL storage volume per well
  • Proteins and DNA will not adhere to PP, allowing for complete sample recovery.
  • Natural color
  • Nonsterile and sterile available

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
50 Cases
96-well Storage Microplate
1300 μL
1000 μL
Round
8 x 12
Natural
Polypropylene
96
Sterile
For storage of low-binding characteristics of polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers