missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Flat Bottom Chimney Style Non-binding Microplates
To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Flat Bottom Chimney Style Non-binding Microplates are free of detectable DNase, RNase, human DNA.
Brand: Greiner Bio-One 655900
This item is not returnable.
View return policy
Description
- Reduces non-specific biomolecule binding (proteins, DNA, RNA) for biochemical assays
- Stable under normal assay conditions without degradation or leaching
- Improves signal-to-noise-ratio
- Manufactured under DIN ISO 9001 guidel
Service Plan
No
Services
No
Specifications
No | |
Without Lid | |
Non Binding | |
Polystyrene | |
Non-sterile | |
392 μL | |
10 | |
Black |
Standard | |
8 x 12 | |
Black | |
96 | |
Non-treated | |
Flat Bottom (F-well) | |
40/Cs. | |
127.8 x 85.5 x 14.4 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One 96-well Flat Bottom Chimney Style Non-binding Microplates > Black
Spot an opportunity for improvement?Share a Content Correction