Learn More
Eppendorf™ 96-Well Flat Bottom Sterile Polypropylene Microplates
GREENER_CHOICE
Brand:  Eppendorf™ 0030602102
242.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10310604
  • £255.00 / Pack of 80
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Standard
White, Clear Bottom
96
Flat Bottom (F-well)
White
8 x 12
Polypropylene
Sterile
80/Pk.
Clear