missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 96-Well Flipper™ Microtube Racks

Protect samples and allow racks to be stacked using this 96-well microtube rack.

Brand:  Thermo Scientific™ 8790

 View more versions of this product

Product Code. 11560115

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Protect samples and allow racks to be stacked using the Thermo Scientific™ 96-Well Flipper™ Microtube Rack. The rack, which is molded with an alphanumeric grid for easy tube identification, holds 0.5mL tubes on one side and 1.5mL tubes on the other.

  • Holds 0.5mL tubes on one side, 1.5mL tubes on the other
  • Molded with an alphanumeric grid for easy tube identification
  • Several colors available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

96-well Flipper Rack
Red
Polypropylene
10 Racks
Flipper Rack
Tubes (2 mL), Tubes (1.5 mL), Tubes (2.0 mL)
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ 96-Well Flipper™ Microtube Racks > Red

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.