missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, Half Area, High Content Imaging, Low Base, Film Bottom Microplate, with Lid
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Brand: Corning™ 4517
Description
High optical-quality film bottom, black microplates are ideal for performing high content cell-based assays using imaging systems. The low base plate allows for unobstructed reading of all wells even when using high numerical aperture objectives. The film bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput and is ideal for cell growth.The microplate and film are manufactured from cyclic olefin copolymer (COC), which has excellent optical
- 96-Well, half area, high content imaging, low base, film bottom microplate, with lid
- COC material allows for broad chemical resistance (including DMSO) and high mechanical stability.
- Ultra-clear film with 127 μm thickness is well suited for imaging micropscopy.
- Inter- and intra-well film bottom flatness within 50 μm and 10 μm, respectively, optimized for high content applications
- Low auto-fluorescence and birefringence
Specifications
Standard | |
0.16 cm2 | |
Black, Clear Bottom | |
96 | |
Sterile | |
Round, Flat Bottom (F-well) | |
20/Pk. | |
Clear |
With Lid | |
8 x 12 | |
Cycloolefin Copolymer | |
50 μL | |
Tissue Culture | |
20/Cs. | |
Low Base |