Learn More
Thermo Scientific™ 96-Well Microtiter™ Microplates
Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Brand: Thermo Scientific™ 2605
81.75 GBP valid until 2025-03-14
Use promo code "24150" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
- For high-throughput screening and research
- Clear plates for colorimetric applications
- Standard footprint, 128 × 86mm
- Available as both polystyrene square wells and vinyl square wells
- Vinyl Microtiter microplates are used for complement fixation, agglutination, and blood typing
- Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing, and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.

Specifications
Standard | |
8 x 12 | |
Microtiter | |
Microplate | |
280 μL | |
Non-sterile | |
Conical Bottom (V-well) | |
10 |
Without Lid | |
50 Cases, 10 Pk. | |
Clear | |
Polystyrene | |
96 | |
Non-treated | |
50 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Handle with Precision
Save up to 92% on liquid handling productsPromo Code: 24150Expires: 2025-03-14
-
Multiple Manufacturers
New Offer Shop Now
-
Multiple Manufacturers
More Discovery, Less Cost
Save on thousands of eligible products.Promo Code: 24111Expires: 2025-03-14
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.