Learn More
Thermo Scientific™ 96-Well Microtiter™ Microplates
Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Brand: Thermo Scientific™ 2205
105.15 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
Description
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Features:
- Use for high-throughput screening and research
- Clear plates for colorimetric applications
- Standard footprint, 128 × 86mm
- Available as both polystyrene square wells and vinyl square wells
- Vinyl Microtiter microplates are used for complement fixation, agglutination and blood typing
- Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
Specifications
Hydrophobic | |
Without Lid | |
50 Cases, 10 Pk. | |
Clear | |
Polystyrene | |
96 | |
Non-treated | |
50 | |
50/Pk. |
Standard | |
8 x 12 | |
Microtiter | |
Microplate | |
400 μL | |
Non-sterile | |
Round Bottom (U-well) | |
10 | |
U96 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.