missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 96-Well Microtiter™ Microplates
Click to view available options
Material:
Polystyrene
Vinyl
Well Shape:
Conical Bottom (V-well)
Curved Edges, Flat Bottom (C-well)
Flat Bottom (F-well)
Round Bottom (U-well)
Volume (Metric) Well:
280 μL
330 μL
400 μL
Lid:
With Lid
Without Lid
Unit Size:
Pack of 100
Pack of 50
Description
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
- For high-throughput screening and research
- Clear plates for colorimetric applications
- Standard footprint, 128 × 86mm
- Available as both polystyrene square wells and vinyl square wells
- Vinyl Microtiter microplates are used for complement fixation, agglutination, and blood typing
- Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing, and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Packaging | 100 Pk. |
| Product Line | Microtiter |
| Color | Clear |
| Product Type | Microplate |
| Material | Vinyl |
| Volume (Metric) Well | 330 μL |
| No. of Wells | 96 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction