Learn More
BTX™ HT 100 Plate Handler, Manual Track Switching
Designed to work with 96 well plates. BTX™ Harvard Apparatus 96 Well Plate Handler helps to create a safe and reliable working area for processing electroporation plates.
Brand:  BTX™ 450400
Product Code. 15477300
  • £4610.00 / Each
Explore more special offers
Description
Description
  • Quickly optimize for better gene delivery
  • Designed to work with 96 well plates
  • Pulse switching technology integrated into package and gold plated contacts to mate with electroporation plate
  • With an adapter to accommodate 25 well plates as well with our ECM 630 generator
  • Prosess up to 96 wells in seconds
Specifications
Specifications
25-well, 96-well
ECM630, ECM830, Gemini X2 electroporation systems
Electroporation accessory
HT-100 Microplate Handler
Pin
1 to 99
5°C to 40°C
20 to 80%
Banana Cables
Exponential Decay Wave, Square Wave
High-Throughput Electroporation
BTX Electroporation
3000 VDC, 250 VAC
LCD
Manual track switching, Connectors for standard pulse generators
10 μs to 10 s
1 Ea.
2 mm, 4 mm
Digital User
Yes