Learn More
Thermo Scientific™ WebSeal 96-Well Non-Coated Plastic Microplates
For any standard application or assay, you can be sure of excellent solvent resistance and low background noise with these high quality polypropylene 96-well microplates.
£309.52 - £804.00
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Product Line | Webseal |
Product Type | Microplate |
Material | Polypropylene |
Product Code | Brand | Total Volume (Metric) Well | No. per Pack | Well Shape | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Total Volume (Metric) Well | No. per Pack | Well Shape | Price | Quantity & Availability | ||||||
15884304
|
Thermo Scientific™
60180-P129 |
145 μL | 100 Microplates | Conical Bottom (V-well) |
£663.30
Pack of 100 |
In Stock
Log in to see stock. |
||||||
15851435
|
Thermo Scientific™
60180-P127 |
252 μL | 100 Microplates | Conical Bottom (V-well) |
£804.00
Pack of 100 |
In Stock
Log in to see stock. |
||||||
15836261
|
Thermo Scientific™
60180-P154 |
1100 μL | 50 Microplates | Round Bottom (U-well) |
£340.70
Pack of 50 |
In Stock
Log in to see stock. |
||||||
17493193
|
Thermo Scientific™
60180-P124 |
2000 μL | 50 Microplates | Round Bottom (U-well) |
£309.52
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15172699
|
Thermo Scientific™
60180-P134 |
2000 μL | 60 Microplates | Round Bottom (U-well) | N/A | N/A | N/A | |||||
Description
Whatever your standard or routine application or assay, you can be sure of excellent solvent resistance and low background noise with Thermo Scientific™ WebSeal 96-Well Non-Coated Plastic Microplates. Made of high-quality, GC-tested polypropylene, these microplates are ideal for:
- Pharmaceutical and industrial QA/QC
- High-throughput screening (HTS)
- Sample collection and storage
- Combinatorial chemistry
- Liquid phase assay
Chemically and Thermally Resistant Polypropylene
- Broad chemical compatibility with alcohols, acetonitrile and other common HPLC solvents
- Will tolerate temperatures from −80°C to 121°C
- GC-tested to ensure low extractables
Broad Instrument Compatibility
- SBS and ANSI standard footprint design
Well Formats to Suit Every Application
- Shallow well and deep well formats
- U- and V- bottom wells ensure optimal sample recovery
Specifications
Standard | |
Webseal | |
Polypropylene | |
Non-coated |
8 x 12 | |
Microplate | |
Non-sterile |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.