missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-Well Protein LoBind Deep Well Plates
GREENER_CHOICEBIOPHARMA

Use these plates in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results. Eppendorf™ Deepwell Protein LoBind Plates recover significantly more protein for downstream analyses and enzymes remain active.

Brand:  Eppendorf™ 0030504216

287.36 GBP valid until 2024-11-01
Use promo code "21693" to get your promotional price.


 View more versions of this product

Product Code. 10115602

  • £318.00 / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • LoBind material guarantees maximum sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: of human DNA, DNAase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Unique OptiTrack™ matrix: 30% faster sample identification and fewer pipetting errors
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface guarantee reliable sealing in plates
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yes
Quantitative Protein Analysis
20 Plates (5 Bags x 4 Plates)
Polycarbonate
96
Yes
121°C
Deep well plates, 96 Wells
Standard
8 x 12
Yellow
1000 μL
Conical Bottom (V-well)
For PCR techniques
20 Pk.
20 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers