missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates

missing translation for 'orderingAttributeHoverText'
Temperature Range (Metric):
-20°C to +120°C
-20°C to +130°C
-20°C to +55°C
Well Shape:
Round
Square
Fit for Purpose:
PCR Ready
missing translation for 'unitSize'
Pack of 50
Description
Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and piercable formats.
Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either piercable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.
Highlights
- Certified RNAse, DNAse, PCR Inhibitor, and human DNA free
- Round or square capped mats for sealing 96-well storage plates
- Alphanumeric grid-referencing
- Molded from a proprietary thermoplastic elastomer for a flexible design
- Suitable for long term storage
- Available in autoclavable or piercable formats
Pierceable version:
- Solvent resistant (including DMSO, Ethanol, and Isopropanol)
- Suitable for Gamma Irradiation
- Requires 6N for piercing
- Temperature Range: −20°C to +55°C
Autoclavable version:
- Suitable for Gamma Irradiation
- Temperature Range: −20°C to +130°C
Hazardous: No
Storage Condition: Ambient
Specifications
Specifications
| Color | Natural |
| Solvent Or Alcohol Resistant | Alcohol Resistant, Solvent Resistant |
| Description | CapMat for 96-well Polypropylene Sample Processing & Storage Plates |
| Certifications/Compliance | Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant |
| Material | TPE |
| Sterility | Cleanroom |
| Autoclavable | Autoclavable |
| No. of Wells | 96 |
| No. per Case | 50 |
| No. per Pack | 10 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction