Learn More
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates
Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.
Brand: Eppendorf™ 0030129660
169.00 GBP valid until 2024-12-31
Use promo code "23292" to get your promotional price.
Additional Details : Weight : 0.85000kg
Description
- LoBind material guarantees maximum sample recovery for improved assay results.
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
Specifications
No Barcode | |
PCR Clean | |
Standard | |
8 x 12 | |
25 Plates (5 Bags x 5 Plates) | |
Green | |
PCR Plate | |
Polypropylene, Polycarbonate | |
96 | |
Conical Bottom (V-well) | |
Yes | |
25 | |
PCR Clean (Non-sterile) | |
Yes |
Low DNA Binding | |
Yes | |
With Lid | |
Autoclavable | |
Eppendorf | |
Single-use | |
Skirted | |
150 μL | |
Non-sterile | |
Real Time PCR | |
6000 x g | |
Yes | |
25 Pk. | |
Colorless |