Learn More
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates
GREENER_CHOICE
Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.
Brand:  Eppendorf™ 0030129555
 View more versions of this product
Product Code. 15861525
  • £1701.78 / Pack of 300
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • LoBind material guarantees maximum sample recovery for improved assay results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Includes Barcode
PCR Clean
Standard
8 x 12
300 Plates (10 Bags x 30 Plates)
Green
PCR Plate
Polypropylene, Polycarbonate
96
Conical Bottom (V-well)
Yes
300 Pk.
PCR Clean (Non-sterile)
Yes
Low DNA Binding
Yes
With Lid
Autoclavable
Eppendorf
Single-use
Skirted
150 μL
Non-sterile
Real Time PCR
6000 x g
Yes
300 Pk.
Colorless