missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates
GREENER_CHOICE

Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors. Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates feature a one-piece design-combining polycarbonate and polypropylene for optimum performance.

Brand:  Eppendorf™ 0030128613

121.34 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.


 View more versions of this product

Product Code. 10777294

  • £138.55 / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors. Eppendorf™ 96-Well twin.tec™ Semi-Skirted Conical Bottom PCR Plates feature a one-piece design-combining polycarbonate and polypropylene for optimum performance.

  • Improved well-to-well tolerance
  • Semi-skirt for labelling
  • Extremely thin-walled for optimum heat transfer into the sample
  • Cut-away corner and alphanumeric grid-referencing
  • Raised well rims for effective sealing and reduced cross-contamination risk
  • 250μL well volume (when used with cap strips)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yes
Quantitative protein analysis
Eppendorf
PCR Microplate
Polycarbonate, Polypropylene
96
Yes
121°C
121°C
Standard
8 x 12
Red, Clear
Semi
250 μL
Conical Bottom (V-well)
For PCR techniques
96 Wells, Semi-Skirted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.