missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 9790 Visor Adaptors
For attaching visors to safety helmets.
£6.95 - £21.60
Specifications
Color | Black |
---|---|
For Use With (Application) | Attaching Visors to Safety Helmets |
Product Type | Visor Adapter |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17239211
|
Uvex™
9790078 |
Pheos K2H Magnet (2600205) |
£21.60
1 pair |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
17259201
|
Uvex™
9790067 |
Pheos Alpine |
£6.95
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
- Adapter for attaching visors to Uvex™ safety helmets
Specifications
Black | |
Attaching Visors to Safety Helmets | |
Visor Adapter |