Learn More
Uvex™ 9906 Pheos Replacement Visor Lens
Replacement visor lens for pheos series safety helmets is compatible with model 9906.002 (mechanical lever arms), or model 9906.003 (self-locking magnetic catches).
Brand: Uvex™ 9906013
22.86 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Additional Details : Weight : 0.12000kg
Description
- Replacement lens for uvex pheos model 9906.002 (mechanical lever arms) or model 9906.003 (self-locking magnetic catches)
- Lightweight, robust face shield for uvex pheos industrial safety helmet ES-WR / BS-WR and uvex pheos alpine (visor adapter 9790.067 required)
- Labeling: 2C-1.2 W1 B 9 KN
- Features supravision excellence coating
Supravision Excellence Coating
- Inside: Anti-fog
- Outside: Scratch-resistant and chemical-resistant
- Easy to clean, and less susceptible to soiling
Specifications
EN 166:2001, EN 170:2002 | |
Pheos 9906.002, 9906.003 | |
Clear | |
Polycarbonate |