missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 9970 Cleaning Station and Accessories
3.82 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Product Type:
Cleaning Station
Plastic Pump
Description
- Cleaning Station is easy to mount and refill
- For use with all Uvex™ Safety Spectacles
Specifications
Certifications/Compliance | EN 166:2001 |
Color | Transparent |
Product Type | Plastic Pump |
For Use With (Equipment) | Cleaning Fluid 9972103 |
Disposable | Reusable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'