missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 9972 Microfiber Cloth
5.38 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Black
Description
- Black microfiber cleaning cloth
- For scratch-free cleaning of Uvex™ safety glasses and lenses
Specifications
Certifications/Compliance | EN 166:2001 |
Color | Black |
For Use With (Equipment) | Uvex™ Safety Spectacles |
For Use With (Application) | Scratch-Free Cleaning |
Material | Microfiber |
Disposable | Reusable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'