missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 366
Potent and selective G9a/GLP inhibitor
£191.00 - £733.00
Specifications
Purity | >98% |
---|---|
Product Type | A 366 |
Molecular Weight (g/mol) | 329.44 |
Inhibitors | A 366 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713968
|
Tocris Bioscience™
5163/10 |
10 mg |
£191.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18770298
|
Tocris Bioscience™
5163/50 |
50 mg |
£733.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective G9a/GLP histone lysine methyltransferase inhibitor (IC50 = 3.3 nM). Exhibits >1000-fold selectivity for G9a/GLP over 21 other methyltransferases. Decreases levels of lysine 9 dimethylation on histone H3 (H3K9Me2) in PC3 cells.Specifications
>98% | |
A 366 | |
329.44 | |
A 366 |
For Research Use Only