missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A 784168, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent TRPV1 antagonist (IC50 = 25 nM for inhibition of TRPV1 activation by 50 nM capsaicin). Displays no activity against a range of receptors, including TRPA1, GABA, opioid, and purinergic receptors.
Specifications
Quantity | 10 mg |
Formula Weight | 479.4 |
Percent Purity | >99% |
Chemical Name or Material | A 784168 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'