missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A 867744, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Positive allosteric modulator of α7 nAChRs (IC50 values are 0.98 and 1.12 μM for human and rat α7 receptor ACh-evoked currents respectively, in X. laevis oocytes). Displays no activity at 5-HT3A, α3β4 or α4β2 nAChRs. Brain penetrant.
Specifications
Quantity | 10 mg |
Formula Weight | 402.89 |
Percent Purity | >97% |
Chemical Name or Material | A 867744 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'